missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoposide, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Etoposide is a DNA topoisomerase II inhibitor (IC50 = 59.2 μM). Anticancer. Also enhances adeno-associated virus (AAV) transduction of HeLa cells.
Specifications
Specifications
CAS | 33419-42-0 |
Molecular Formula | C29H32O13 |
MDL Number | MFCD00869325,MFCD00869325 |
Quantity | 100 mg |
Synonym | vjjpusntgommgy-nzlmilqcsa |
InChI Key | VJJPUSNTGOMMGY-MRVIYFEKSA-N |
SMILES | COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12 |
IUPAC Name | (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one |
Molecular Weight (g/mol) | 588.56 |
PubChem CID | 50936917 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction