missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etrolizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541929
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Etrolizumab has been used in trials studying the treatment of Crohn Disease and Ulcerative Colitis.Specifications
Etrolizumab | |
Recombinant Monoclonal | |
Unconjugated | |
PRO145223; RG7413; rhuMAb Beta7 | |
Human ITGA4_ITGB7 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1.41 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |