missing translation for 'onlineSavingsMsg'
Learn More

Eugenol, 99%, Thermo Scientific Chemicals

Product Code. 10550591
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
Synonym eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
PubChem CID 3314
ChEBI CHEBI:4917
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

Melting Point -12°C to 10°C
Color Yellow
Density 1.0600g/mL
Boiling Point 254.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (H2C=CHCH2)C6H3(OCH3)OH
Refractive Index 1.5400 to 1.5420
Quantity 500 g
Beilstein 06,961
Merck Index 15,394
Specific Gravity 1.06
Solubility Information Solubility in water: 2.46g/L (25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
IUPAC Name 2-methoxy-4-(prop-2-en-1-yl)phenol
Formula Weight 164.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Eugenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Eugenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.