missing translation for 'onlineSavingsMsg'
Learn More

MULTIROIR Euronorm ESD Container

Simplified cleaning thanks to smooth internal walls designed for the storage and transport of electronic products

Brand:  MULTIROIR AESDEG43/32HG

 View more versions of this product

Product Code. 17236870

  • £23.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Euronorm ESD container
  • Material: polypropylene 2 integrated ergonomic handles
  • Simplified cleaning thanks to smooth internal walls
  • Specially designed for the storage and transport of electronic products
Specifications

Specifications

Storage Box
Gray
BP4320 K
Polypropylene
Rectangular
400 x 300 x 320 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers