missing translation for 'onlineSavingsMsg'
Learn More
Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11375749

Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11375749
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11375749

Brand: Thermo Scientific Alfa Aesar 010175.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
Synonym europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oilShow MoreShow Less
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Color Gray
UN Number UN3208
Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.82g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Europium foil, 0.25mm (0.01 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ02K054 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number010175.FI,010175.FF
Lot NumberT11K039 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog Number010175.FI,010175.FF
Lot NumberQ07H036 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number010175.FI,010175.FF
Lot NumberD02N17 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010175.FI,010175.FF
Lot NumberZ21D012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010175.FI,010175.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.