Learn More
Europium(II) bromide, 99.99% (metals basis), Thermo Scientific Chemicals
£62.71 - £800.99
Chemical Identifiers
CAS | 13780-48-8 |
---|---|
Molecular Formula | Br2Eu |
Molecular Weight (g/mol) | 311.772 |
MDL Number | MFCD01076548 |
InChI Key | PJVPGMOCWCUQHP-UHFFFAOYSA-L |
Synonym | europium dibromide, europium ii bromide, europium ii ; bromide, acmc-1bp16, europium ii bromide trace rare earth metals basis 1gShow More |
PubChem CID | 53249294 |
IUPAC Name | europium(2+);dibromide |
SMILES | [Br-].[Br-].[Eu+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480720
|
Thermo Scientific Alfa Aesar
044431.03 |
1 g |
£62.71
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490720
|
Thermo Scientific Alfa Aesar
044431.06 |
5 g |
£215.07
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400730
|
Thermo Scientific Alfa Aesar
044431.14 |
25 g |
£800.99
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Europium(II) bromide is used in water treatment, chemical analysis and in certain crystal growth applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13780-48-8 | |
311.772 | |
PJVPGMOCWCUQHP-UHFFFAOYSA-L | |
53249294 | |
[Br-].[Br-].[Eu+2] |
Br2Eu | |
MFCD01076548 | |
europium dibromide, europium ii bromide, europium ii ; bromide, acmc-1bp16, europium ii bromide trace rare earth metals basis 1gShow More | |
europium(2+);dibromide |
Specifications
13780-48-8 | |
Powder | |
Br2Eu | |
europium dibromide, europium ii bromide, europium ii ; bromide, acmc-1bp16, europium ii bromide trace rare earth metals basis 1g | |
PJVPGMOCWCUQHP-UHFFFAOYSA-L | |
europium(2+);dibromide | |
53249294 | |
Odorless | |
Europium(II) bromide |
White | |
99.99% (Metals basis) | |
MFCD01076548 | |
Soluble in water. | |
[Br-].[Br-].[Eu+2] | |
311.772 | |
311.77 | |
Hygroscopic |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 13780-48-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.