missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11356658
Click to view available options
Quantity:
2 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181266-82-0
Molecular Formula C15H21EuO6
Molecular Weight (g/mol) 449.29
MDL Number MFCD00192170
InChI Key YMTKZYSHNNXTIJ-UHFFFAOYSA-N
Synonym europium iii 2,4-pentanedionate hydrate reo
PubChem CID 131675702
SMILES [Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 140°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
IUPAC Name europium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
Formula Weight 449.29 (Anhydrous)
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Europium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'