missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11360118

Europium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11360118
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11360118

Brand: Thermo Scientific Alfa Aesar 042878.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Europium(III) acetate hydrate is used as fluorescent powder and as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62667-64-5
Molecular Formula C6H9EuO6
Molecular Weight (g/mol) 329.10
MDL Number MFCD00150115
InChI Key LNYNHRRKSYMFHF-UHFFFAOYSA-K
Synonym europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
PubChem CID 16212055
SMILES [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
IUPAC Name europium(3+) triacetate
Formula Weight 329.10 (Anhydrous)
Sensitivity Hygroscopic
Appearance White crystalline powder
Chemical Name or Material Europium(III) acetate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR10K027 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number042878.06,042878.14
Lot NumberY01F021 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number042878.06,042878.14
Lot NumberT24B051 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042878.06,042878.14
Lot NumberJ22W024 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042878.06,042878.14
Lot NumberC02W027 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042878.06,042878.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.