missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh, Thermo Scientific Chemicals

Catalog Number 10616232
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
13759-92-7
Cl3Eu·6H2O
366.4
MFCD00149701
13759-92-7
Cl3Eu·6H2O
366.4
MFCD00149701

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 850.0°C
Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula EuCl3·6H2O
Merck Index 10, 3854
Solubility Information Solubility in water: soluble
Formula Weight 366.4
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.89
Density 4.8900g/mL
Chemical Name or Material Europium(III) chloride hexahydrate, 99.9%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'