missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride, ultra dry, 99.99% (REO)

Product Code. 11398607
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11398607

Brand: Thermo Scientific Alfa Aesar 035738.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-76-0 | Cl3Eu | 258.31 g/mol

Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-76-0
Molecular Formula Cl3Eu
Molecular Weight (g/mol) 258.31
MDL Number MFCD00010994
InChI Key NNMXSTWQJRPBJZ-UHFFFAOYSA-K
Synonym acmc-20akvq, europium 3+ trichloride, europium 3+ ion trichloride, europium iii chloride, ultra dry reo, europium chloride, anhydrous, europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
PubChem CID 9859933
SMILES [Cl-].[Cl-].[Cl-].[Eu+3]

Specifications

Melting Point 850°C
Physical Form Powder
Quantity 5 g
Merck Index 14,3904
Solubility Information Soluble in water; Soluble in dilute hydrochloric acid,acetone and alcohol; Slightly soluble in tetrahydrofuran.
IUPAC Name europium(3+);trichloride
Formula Weight 258.32
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.