Learn More
Europium(III) fluoride, anhydrous, 99.5% (REO), Thermo Scientific™
£36.97 - £441.00
Chemical Identifiers
CAS | 13765-25-8 |
---|---|
Molecular Formula | EuF3 |
Molecular Weight (g/mol) | 208.96 |
MDL Number | MFCD00010998 |
InChI Key | HPNURIVGONRLQI-UHFFFAOYSA-K |
Synonym | europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1gShow More |
IUPAC Name | europium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[Eu+3] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13765-25-8 | |
208.96 | |
HPNURIVGONRLQI-UHFFFAOYSA-K | |
europium(3+) trifluoride |
EuF3 | |
MFCD00010998 | |
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1gShow More | |
[F-].[F-].[F-].[Eu+3] |
Specifications
>2280°C | |
13765-25-8 | |
EuF3 | |
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g | |
[F-].[F-].[F-].[Eu+3] | |
208.96 | |
99.5% | |
(REO) |
1390°C | |
Powder | |
MFCD00010998 | |
HPNURIVGONRLQI-UHFFFAOYSA-K | |
europium(3+) trifluoride | |
208.96 | |
Hygroscopic | |
Europium (III) fluoride, Anhydrous |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 237-368-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.