missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11336918
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
13765-25-8
EuF3
208.96
MFCD00010998
HPNURIVGONRLQI-UHFFFAOYSA-K
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
europium(3+) trifluoride
[F-].[F-].[F-].[Eu+3]
This item is not returnable. View return policy
13765-25-8
EuF3
208.96
MFCD00010998
HPNURIVGONRLQI-UHFFFAOYSA-K
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
europium(3+) trifluoride
[F-].[F-].[F-].[Eu+3]

Europium(III) fluoride is used as a catalyst in the preparation of novel mixed-metal fluorides. It finds application in studies related to fluoride glasses. It is also used as a laboratory reagent. Further, it is used in optical glasses and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point >2280°C
Melting Point 1390°C
Physical Form Crystalline
Quantity 2 g
Solubility Information Slightly soluble in strong mineral acids. Insoluble in water.
Formula Weight 208.96
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'