missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) fluoride, anhydrous, REacton™, 99.98% (REO), Thermo Scientific Chemicals Product Code.: 11331049

Europium(III) fluoride, anhydrous, REacton™, 99.98% (REO), Thermo Scientific Chemicals

Product Code. 11331049
5 g
Click to view available options
Quantity:
2 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11331049

Brand: Thermo Scientific Alfa Aesar 011302.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-25-8
Molecular Formula EuF3
Molecular Weight (g/mol) 208.96
MDL Number MFCD00010998
InChI Key HPNURIVGONRLQI-UHFFFAOYSA-K
Synonym europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
IUPAC Name europium(3+) trifluoride
SMILES [F-].[F-].[F-].[Eu+3]

Specifications

Boiling Point >2280°C
Melting Point 1390°C
Physical Form Powder
Quantity 5 g
Formula Weight 208.96
Percent Purity 99.98%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) fluoride, Anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT06D042 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number011302.06,011302.14,011302.04
Lot Number10253633 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number011302.06,011302.14,011302.04
Lot NumberN28I105 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011302.06,011302.14,011302.04
Lot Number22177 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011302.06,011302.14,011302.04
Lot NumberD07K09 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011302.06,011302.14,011302.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) fluoride, anhydrous, REacton™, 99.98% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.