missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

Product Code. 10733391 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10733391

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-53-5 | EuN3O9·6H2O | 446.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 10031-53-5
Physical Form Crystalline Solid
Quantity 5 g
Assay Percent Range 99.90%
Molecular Formula EuN3O9·6H2O
Linear Formula Eu(NO3)3·6H2O
MDL Number MFCD00149700
Synonym europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g
InChI Key JVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3]
IUPAC Name europium(3+);trinitrate;hexahydrate
Molecular Weight (g/mol) 446.07
PubChem CID 202256
Formula Weight 446.07
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Europium(III) nitrate hexahydrate, 99.9%
Show More Show Less
compliance-icons
Product Identifier
  • Europium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.