missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11393438
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

Europium(III) oxide is used for vacuum deposition, fluorescent lamps, and as an activator for yttrium-based phosphors, manufacture of fluorescent glass, anti-counterfeiting phosphors in Euro banknotes. And it is widely used as a red or blue phosphor in television sets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Physical Form Sintered Lump
Quantity 10 g
Merck Index 14,3904
IUPAC Name europium(3+);oxygen(2-)
Formula Weight 351.92
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'