missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10056333

Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10056333
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10056333

Brand: Thermo Scientific Acros 193020250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Color White to Pink
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99%
Linear Formula Eu2O3
Merck Index 10, 3854
Solubility Information Solubility in water: insoluble
IUPAC Name europium(3+);oxygen(2-)
Formula Weight 351.91
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 7.42
Density 7.4200g/mL
Chemical Name or Material Europium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464165 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number193020050,193020250,193020010
Lot NumberA0454375 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number193020050,193020250,193020010
Lot NumberA0424957 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number193020050,193020250,193020010
Lot NumberA0451349 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number193020050,193020250,193020010
Lot NumberA0438746 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number193020050,193020250,193020010
Product Identifier
  • Europium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.