missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific™

£354.77

Chemical Identifiers

CAS 13537-22-9
Molecular Formula Cl3EuO12
Molecular Weight (g/mol) 450.30
MDL Number MFCD00016076
InChI Key KDBBDPJZIDRZFY-UHFFFAOYSA-K
Synonym europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol.Show MoreShow Less
PubChem CID 11733073
IUPAC Name europium(3+);triperchlorate
SMILES [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410301
SDS View Documents
Thermo Scientific Alfa Aesar
040572.NC
10 g
£354.77
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.