missing translation for 'onlineSavingsMsg'
Learn More
Europium pieces, dendritic, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15421017

Europium pieces, dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15421017
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 15421017

Brand: Thermo Scientific Alfa Aesar 012394.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
Synonym europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Color Gray-Silver
UN Number UN3208
Quantity 5 g
Percent Purity 99.9%
Odor Odorless
Sensitivity Air and moisture sensitive
Form Pieces
Assay (REO)
Chemical Name or Material Europium pieces, Sublimed dendritic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY20K002 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number012394.06
Lot NumberZ23K027 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number012394.06
Lot NumberP25I007 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number012394.06
Lot NumberT08A011 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012394.06
Lot NumberF3635 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012394.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium pieces, dendritic, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.