missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech Handheld Meters Calibration Solution, 1413μS/cm KCl

Periodic calibration with accurate standards is recommended for meters

Brand:  Thermo Scientific™ 01X211207

13.53 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 15284878

  • £14.24 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 1413μS/cm KCl calibration solution
  • Solutions packed in 480mL durable plastic bottles
  • Economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Inorganic Salts
7447-40-7
450 mL
Complex
1413 μS/cm calibration solution
7.0
Eutech Handheld Meters
Calibration Solution
Nil
1 case
1
Liquid
Colorless
102.4°C
Calibration Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Eutech Handheld Meters Calibration Solution, 1413μS/cm KCl >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.