Learn More
Thermo Scientific™ Eutech Handheld Meters Calibration Solution, 84μ S / cm KCl
Brand:  Thermo Scientific™ 01X211241
Product Code. 15264878
  • £14.66 / 480mL
Explore more special offers
Description
Description
  • 84μS/cm KCl calibration solution
  • Solutions packed in 480mL durable plastic bottles
  • Economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
480 mL
Calibration Solution
Odorless
∼7
Laboratory Use
Calibration Solution
Eutech Handheld Meters
1.0 g/cm³ at 25°C
∼100°C