Learn More
Eutech™ 5.0 mS/cm KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Brand:  Thermo Scientific Eutech 01X211272
Product Code. 15263939
  • £14.74 / 480mL
Explore more special offers
Description
Description
Eutech™ 5.0 mS/cm KCl Calibration Solution is used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 480ml durable plastic bottles
  • Economical and convenient to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
480 mL
Calibration Solution
Conductivity Standard
Economical and Convenient to Use