Learn More
Eutech™ Conductivity Calibration Sachets
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Brand:  Thermo Scientific Eutech 01X219901
 View more versions of this product
Product Code. 11543775
  • £76.62 / Each
Explore more special offers
Description
Description
Get quick, accurate, and convenient conductivity measurements with NIST_traceable Eutech™ Conductivity Calibration Sachets. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
  • Conductivity Standards Sachet -20 ml
  • NIST Traceable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1413 μS/cm KCl Conductivity Sachets
Calibration Solution
Field, School, Laboratory
20 x 20 mL
Conductivity Standard