missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ KCl Calibration Solution

For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions

Brand:  Thermo Scientific Eutech 01X211214

 View more versions of this product

Product Code. 15386487

  • £14.24 / 480mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.

Features:

  • Packed in 60mL or 480 ml durable plastic bottles
  • Economical and convenient to use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2764 μS/cm KCl calibration solution
Calibration Solution
Conductivity Meters and Electrodes
Economical and Convenient to Use
480 mL
Conductivity Standard
2764 μS/cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.