Learn More
Eutech™ KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Brand:  Thermo Scientific Eutech 01X608301
 View more versions of this product
Product Code. 17266033
  • £8.81 / 60mL
Explore more special offers
Description
Description
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 60mL or 480 ml durable plastic bottles 
  • Economical and convenient to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1413 μS/cm KCl Calibration Solution
Calibration Solution
1413 μS/cm
60 mL
Conductivity Meters and Electrodes