Learn More
Eutech™ ORP Solutions
Eutech™ offers ORP Standard Solutions for use with ORP Meters.
Brand: Thermo Scientific Eutech 01X081500
Code : 04
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Description
ORP meters can measure both pH and ORP values in various educational, laboratory and industrial applications. Major areas of usage include the treatment of industrial wastes, study of biological systems, oxidation of cyanide, bleaching of pulp, manufacture of bleach and reduction of chromate wastes. The measurement of ORP is also useful in pool water treatment as an indication of sanitation in relation to free chlorine parameter. Eutech™ offers a range of ORP Solutions for use with our ORP Meters.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Pre-treatment solution (475 mV) | |
ORP Meter |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.