Learn More
Thermo Scientific™ Eutech™ PCTestr 35 Multi-Parameter Pocket Tester
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Brand: Thermo Scientific™ 01X441504
212.54 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 04
Additional Details : Weight : 0.18270kg
Description
No need to change sensor every time you switch parameters. The Eutech™ PCTestr 35 Multi-Parameter Pocket Tester measures three parameters (measures pH, Conductivity and Temperature). For general applications: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required. PCSTestr 35 meets requirements for many standard environmental and ASTM test methods. For industrial applications: Use for cooling towers, food processing water testing and process/wastewater testing in metal finishing, photo development, printing and chemical industries. For educational applications: Useful in schools, many laboratory applications and ecology studies.
Product Features
- Full pH rangemeasurement at up to 2-decimal places.
- Low,medium and high Conductivity ranges – testermeasures pure water!
- Up to 5-point pH calibration and 3-point Conductivity calibration.
- Adjustable Temperature Coefficient (0.0 to 0.0%)
- Temperature sensor housed within unique dome-shaped casing for faster ATC response and fastermeasurements.
- ATC/mTC option
- Scroll-menu with four buttons control for stress-free navigation
- Longest-lasting tester in themarket! No tools required to replace sensor.
- 'One-press Parameter Toggle': Switching parameters is just a press of the button.
- Certified IP67 Waterproof – Lightweight tester even floats for easy retrieval.
Specifications
- pH Range: 0.00 to 14.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- pH Calibration Points: Up to 5 (Auto)
- pH Buffer Options: USA, NIST
- Conductivity Range: .. to 1999 μS/cm, 2.00 to 20.00mS/cm
- Resolution: 1 μS/cm, 0.01mS/cm
- Accuracy: ±1% Full Scale
- Conductivity Calibration Points: 2 Auto, 2manual
- Normalization Temperature: 25.0 °C
- Temperature Coefficient: 0.0 to 10.0%/°C
- Auto-Ranging: Yes
- Temperature Range: 0.0 to 50.0 °C / 32.0 to 120.0 °F
- Resolution: 0.1 °C/°F
- Accuracy: ±0.5°C/ ±0.9°F
- Temperature Compensation: ATC /mTC
- Non-Volatilememory: Yes
- Auto-Off: 8.5mins after last key pressed (can be disabled)
- LCD Display: Dual-display LCD (2.1 x 2.7cm)
- Power: 4 X 1.5 V 'A76'micro alkaline batteries
- Battery Life: > 150 hours
- Dimensions; Weight: Boxed: 18.5 cm x 6.5 cm x 5 cm; 200 g