missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ pH 150 Meter Kit
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
£362.76 - £423.80
Specifications
Accuracy (pH) | ±0.01 |
---|---|
Accuracy (Temperature) | ±0.5°C;/±0.9°F |
Calibration | mV: ±200 mV, Temp: Offset in 0.1 increments, ±10°C or ±18°F |
Range (pH) | -2 to 16 |
Resolution (pH) | 0.01 |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15377043
|
Thermo Scientific™
01X651607 |
pH 150 Meter Only: Meter, Grip-Clip™, two AA batteries |
£362.76
Each |
In Stock
Log in to see stock. |
||||||
15387043
|
Thermo Scientific™
01X651608 |
pH 150 Meter & Electrode: Meter with double-junction pH electrode ECFC7252201B and ATC probe PHWPTEM01W, Grip-Clip™, two AA batteries |
£423.80
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
Description
Eutech™ offers a wide range of pH meters and electrodes. pH 150 Meter Kit is an an IP67-rated waterproof pH meter that offers measurement of pH / mV / Temperature.
Features
- Large, three line LCD – View pH or mV measurement with temperature and % slope
- Built-in memory function – Stores up to 150 measurements along with temperature
- Auto buffer recognition – Automatically identifies USA, NIST or DIN buffer sets for rapid calibration
- Power with AA batteries or use optional AC adapter for benchtop measurements
- Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
- Selectable ºF/ºC – Choose units to best fit your application
- Features mV offset – Calibrate to a Redox/ORP standard or use for pH electrode diagnostics
- Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
- % slope and temperature displayed with each measurement
- Built-in stand – Ideal for easy benchtop or long-term use
- Wall-mount option – Ideal to save on bench space
- Includes innovative Grip-Clip™ – Quickly and easily secures the electrode to your beaker or container
- Compatible with pH electrodes having a BNC connector
Specifications
±0.01 | |
mV: ±200 mV, Temp: Offset in 0.1 increments, ±10°C or ±18°F | |
0.01 | |
Up to 5 (USA & NIST), Up to 6 (DIN) | |
14.0°F to 230.0°F | |
LCD, 3-Line Custom | |
150 | |
±2000 |
±0.5°C;/±0.9°F | |
-2 to 16 | |
0.1 mV ±999.9 mV; 1 mV beyond | |
-10°C to +110°C | |
0.1°C/°F | |
Stores up to 150 data sets | |
Two AA batteries (included), >700 hours continuous use; AC adapter (optional) | |
Automatic/Manual (-10°C to 110°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Eutech™ pH 150 Meter Kit