Learn More
Thermo Scientific™ Eutech™ pH 2700 pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Brand: Thermo Scientific™ 01X543902
907.38 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : A0
Additional Details : Weight : 0.65000kg
Description
Oversize screen with large fonts yet compact – the Eutech pH 2700 pH Meter offers an easy to read screen that says more. View pH or ORP readings, with temperature, electrode status, calibration points, date and time all at once!
Features
- Up to 6-point calibration with auto-buffer recognition
- Quick, easy electrode diagnosis withmultiple pH slopes and offset display
- Non-volatilememory holds up to 500 data points – time and date-stamped for GLP compliance
- Calibrate with up to 5 custom pH buffers – use any pH values that are ≥1.0 pH unit apart
- Cal-due alarm – nomore out-dated calibrations!
- Auto-logging function for convenient continuousmonitoring
- Password protection for setup and calibration
Applications
- Educational: Useful formost laboratory, ecological studies and other applications
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing.
Specifications
Product Specification: Description
pH
- Range: -2.000 to 20.000 pH
- Resolution: 0.1 / 0.01 / 0.001 pH
- Accuracy: ±0.002 pH + 1 LSD
- Cal. Points: Up to 6 preset or 5 custom
- Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom
ORP
- Range: ±2000.0mV
- Rel.mV Range: ±2000.0mV
- Resolution: 0.1mV
- Accuracy: ±0.2mV
Temperature
- Range (meter): 0.0 to 100.0 °C / 32.0 to 212.0 °F
- Resolution: 0.1 °C / 0.1 °F
- Accuracy: ±0.3 °C / ±0.5 °F
- Compensation: ATC /mTC
- Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SmPS)
Specifications
Eutech pH 2700 Meter With pH Electrode (ECFG7370101B), ATC Probe (PH5TEMB01P), Integral Electrode Holder, 100/240 VAC Adapter, RS232 cable, & pH Electrode Refill Solution, 60 ml | |
±0.002 | |
0.3°C/0.5°F | |
0.001, 0.01, 0.1 | |
0°C to 100°C | |
Automatic Data Logging |
RS-232 | |
±0.2 | |
-2 to 20 | |
Up to 6 | |
500 with Date and Time Stamp |