missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ pH 2700 pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
£869.09 - £955.14
Specifications
Accuracy (mV) | ±0.2 |
---|---|
Accuracy (pH) | ±0.002 |
Range (pH) | -2 to 20 |
Resolution (pH) | 0.001, 0.01, 0.1 |
Temperature Range (Metric) | 0°C to 100°C |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11573855
|
Thermo Scientific™
01X543901 |
Eutech pH 2700 Meter With Integral Electrode Holder & 100/240 VAC Adapter, RS232 cable |
£869.09
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11583855
|
Thermo Scientific™
01X543902 |
Eutech pH 2700 Meter With pH Electrode (ECFG7370101B), ATC Probe (PH5TEMB01P), Integral Electrode Holder, 100/240 VAC Adapter, RS232 cable, & pH Electrode Refill Solution, 60 ml |
£955.14
Each |
In Stock
Log in to see stock. |
|||||
Description
Oversize screen with large fonts yet compact – the Eutech pH 2700 pH Meter offers an easy to read screen that says more. View pH or ORP readings, with temperature, electrode status, calibration points, date and time all at once!
Features
- Up to 6-point calibration with auto-buffer recognition
- Quick, easy electrode diagnosis with multiple pH slopes and offset display
- Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
- Calibrate with up to 5 custom pH buffers – use any pH values that are ≥1.0 pH unit apart
- Cal-due alarm – no more out-dated calibrations!
- Auto-logging function for convenient continuous monitoring
- Password protection for setup and calibration
- Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom
Applications
- Educational: Useful for most laboratory, ecological studies and other applications
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing.
Specifications
±0.2 | |
-2 to 20 | |
0°C to 100°C | |
Automatic Data Logging |
±0.002 | |
0.001, 0.01, 0.1 | |
500 with Date and Time Stamp |