missing translation for 'onlineSavingsMsg'
Learn More

Eutech pH Buffer Sachets

Get a wide range of Eutech™ pH buffers and solutions for use with pH meters and electrodes

Brand:  Eutech ECBU4710R

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15691707

  • £75.89 / Each
In Stock
to see stock.
Add to basket

Description

Description

pH Buffer Sachets are NIST-traceable sachets for quick, accurate and convenient calibrations. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.

Features:

  • pH Buffer Sachet -20 ml
  • NIST Traceable
Specifications

Specifications

pH buffer sachet set, pH 4.01, 7.00, 10.01, deionized water, 5 x 20 mL each
pH Meter and Electrode
5 x 20 mL
Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers