missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eutech™ pH Buffer Tablets
Click to view available options
pH:
4
7
10
Unit Size:
Pack of 100
Description
Eutech™ pH buffer tablets provide a quick, convenient way to prepare pH buffers from tablets for pH calibrations. Each box contains 100 pH buffer tablets.
Features:
- 10 strips x 10 tablets per box
Specifications
Specifications
| Color | Red |
| For Use With (Application) | pH calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4 |
| Physical Form | Tablet |
| Quantity | 100 Tablets |
| Solution Type | Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction