missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ pH Buffers and Solutions

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes

Brand:  Thermo Scientific Eutech 01X211201

 View more versions of this product

Product Code. 15302811

  • £16.20 / 480mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications

Specifications

Red
pH Meter and Electrode
480 mL
pH 4.01 Buffer Solution
4.01
Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers