Learn More
Eutech™ pH Buffers and Solutions
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
£14.66 - £55.60
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10759503
|
Thermo Scientific Eutech
01X211211 |
pH 1.68 Buffer Solution | 480 mL |
£14.66
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15302811
|
Thermo Scientific Eutech
01X211201 |
pH 4.01 Buffer Solution | 480 mL |
£16.20
480mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15332811
|
Thermo Scientific Eutech
01X211202 |
pH 7.00 Buffer Solution | 480 mL |
£20.00
480mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15303361
|
Thermo Scientific Eutech
01X211203 |
pH 10.01 Buffer Solution | 480 mL |
£20.09
480mL |
In Stock
Log in to see stock. |
||||
10616793
|
Thermo Scientific Eutech
01X211203 |
pH 10.01 Buffer Solution | 480 mL | N/A | |||||
15611717
|
Thermo Scientific
01X638901 |
pH buffer kit, pH 4, 7, 10 clear buffer solutions each. | 480 mL | N/A | |||||
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
pH Meter and Electrode | |
480 mL |
Safety and Handling
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : potassium tetraoxalate dihydrate