Learn More
Thermo Scientific™ Eutech™ pH Electrode Storage Solution
Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes
Brand: Thermo Scientific™ 01X211206
Code : 04
Additional Details : Weight : 0.53820kg
Description
The measure of a liquid’s acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Storage solution for pH electrodes | |
EutechTM pH Electrode | |
pH Electrode Storage Solution |