missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Power Adapter for Temp 300/360 series meters

The Eutech™ Power Adapter provides you with the power you need for your Eutech™ meters.

Brand:  Thermo Scientific™ 60X030134

Product Code. 11503885

  • £43.45 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Eutech™ Power Adapter provides you with the power you need for your Eutech™ meters. The 100/240 VAC Power Adapter is used for Eutech™ 300 series meters.

Features:

  • 9 V, 6 W
  • 100/240 VAC
  • Used With: Temp 300/360 series meters
Specifications

Specifications

100/240 VAC Power Adapter, 9V, 6W, CNT-VE
Adapter
9 V
Temp 300/360 series meters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.