Learn More
Eutech™ Refilling Reference Electrolyte (Double Junction pH electrode)
Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes
Brand: Thermo Scientific Eutech 01X211218
Code : 04
Additional Details : CAS Number : 7447-40-7 Weight : 0.50000kg
Description
The measure of a liquid’s acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Saturated Potassium Chloride for Double Junction pH Electrodes | |
480 mL | |
Laboratory Use |
Refilling Reference Electrolyte | |
Calibration Solution |