Learn More
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions
Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.
Brand: Thermo Scientific™ ECNACL45PPT
This item is not returnable.
View return policy
Description
Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Description:
- ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
- ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
- ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
Specifications
45 ppt NaCl Calibration Solution |