Learn More
Thermo Scientific™ Eutech™ Sensor for ECTestr11
Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
Brand:  Thermo Scientific™ 01X229714
Product Code. 10475982
  • £65.52 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eutech™ offers meters that allow the direct reading of TDS values. Eutech™ Sensor for ECTestr11 is a replacement sensor for waterproof ECTestr11, Cup Type. Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
General applications:
  • water and wastewater treatment
  • boiler blow-down
  • electroplating rinse tanks
  • drinking water
  • hydroponics
  • printing industry
  • aquaculture, aquariums, fish farms
  • swimming pools and spas
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sensor
ECTestr11
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Eutech™ Sensor for ECTestr11 >