Learn More
Thermo Scientific™ Eutech™ Sensor for pHTestr10, 20, 30
Optimize your Eutech™ pH meters with these accessories and replacement parts.
Brand:  Thermo Scientific™ 01X106709
Code : 04
Additional Details : Weight : 0.04000kg
Product Code. 11830303
  • £76.33 / Each
In Stock
to see stock.
Add to basket
Description
Description
Optimize your Eutech™ pH meters with these accessories and replacement parts. Eutech™ Replacement Sensor is for waterproof pHTestr10, 20, 30.
Replacement sensor for pHmeters:
  • pHTestr 10
  • pHTestr 20
  • pHTestr 30
Specifications
Specifications
pH Sensor