Learn More
Thermo Scientific™ Eutech™ Sensor for TDSTestr11
Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
Brand: Thermo Scientific™ 01X229713
Code : 04
Additional Details : Weight : 0.01000kg
Description
Eutech™ offers meters that allow the direct reading of TDS values. Eutech™ Sensor for TDSTestr11 is a replacement sensor for TDSTestr11 series, dip type. Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
General applications:
- water and wastewater treatment
- boiler blow-down
- electroplating rinse tanks
- drinking water
- hydroponics
- printing industry
- aquaculture, aquariums, fish farms
- swimming pools and spas
Specifications
Sensor | |
TDSTestr11 |