Learn More
Thermo Scientific™ Eutech TN-100 Turbidimeter Calibration Kit
Simple, display-prompted push-button calibration
Brand:  Thermo Scientific™ 01X621001
222.36 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Product Code. 11786384
  • £261.90 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 60mL bottles of U.S. EPA-approved, non-formazin standards
  • Stable and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration Kit Set
0.02 NTU, 20.0 NTU, 100 NTU, 800 NTU
Standard solutions; 4 vials of 60mL each
U.S. EPA-approved non-formazin standards