Learn More
Thermo Scientific™ Eutech™ pH 6+ Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
£333.01 - £381.20
Specifications
Accuracy (mV) | ± 1 mV |
---|---|
Accuracy (pH) | ±0.01 |
Battery Type | 4 AAA x 1.4 V |
Calibration Points | Up to 5 |
Display Type | LCD |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11549164
|
Thermo Scientific™
01X245025 |
Eutech pH 6+ pH Meter with ATC Electrode ECPHTEM01P, Rubber Armour/Stand |
£333.01
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11569164
|
Thermo Scientific™
01X245028 |
Eutech pH 6+ pH Meter with ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised Rinse Water -60 ml Each), Rubber Armour/Stand |
£381.20
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. The Eutech™ pH 6+ Meter is an ideal choice for pH applications from the wide range of pH meters that Eutech™ offers.
Features:
- Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Hold Function momentarily freezes reading for easy viewing and recording
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic with message codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
- pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
Specifications
± 1 mV | |
4 AAA x 1.4 V | |
LCD | |
-1000 to 1000 mV | |
0.01 | |
Auto Power-Off after 17 minutes; Hold & Self-diagnostic messages | |
±0.05°C | |
0.1 °C |
±0.01 | |
Up to 5 | |
Acidity, Alkalinity, Conductivity, Temperature, pH | |
0.00 to 14.00 | |
0°C to 100.0°C | |
1 mV | |
Automatic / Manual (0 to 100 °C) |