Learn More
Thermo Scientific™ Eutech™ X5 Membranes and O-rings for Dissolved Oxygen Probes
Optimize your Eutech™ DO probes with these accessories and replacement parts.
£35.71 - £64.71
Specifications
For Use With (Equipment) | Dissolved Oxygen Probe |
---|---|
Type | X5 Membrane and O-Ring |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11723619
|
Thermo Scientific™
01X241603 |
Membrane & O-ring (pack of 5) |
£64.71
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
11569054
|
Thermo Scientific™
15X241502 |
Membrane installation tool |
£35.71
Each |
In Stock
Log in to see stock. |
|||||
Description
Optimize your Eutech™ DO probes with these accessories and replacement parts.
- Membrane/O-ring Replacement is an optional procedure
- It is recommended only experienced service personnel can perform this procedure.
- It should only be performed if you have new membrane and O-ring.
Specifications
Dissolved Oxygen Probe | |
X5 Membrane and O-Ring |