Learn More
Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids
Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!
Brand: Fisherbrand™
Additional Details : Weight : 0.63500kg
Includes: removable lid.
Description
- Sweat-proof ethylene vinyl acetate will not leak
- Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
- Bases and lids feature glove-friendly handles
- Cleanup is quick and easy with molded pour spouts
Specifications
Ice, dry ice, liquid nitrogen, alcohol, and saline solutions | |
Bucket with Lid | |
Cryogenic Storage Box | |
Purple | |
Not Autoclavable |
Removable lid | |
EVA | |
1 | |
2.5 L |