Learn More
Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids
Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!
Marca: Fisherbrand™
Detalles adicionales : Weight : 0.95250kg
Includes: removable lid.
Descripción
- Sweat-proof ethylene vinyl acetate will not leak
- Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
- Bases and lids feature glove-friendly handles
- Cleanup is quick and easy with molded pour spouts
Especificaciones
Ice, dry ice, liquid nitrogen, alcohol, and saline solutions | |
Pan with Lid | |
Cryogenic Storage Box | |
Green | |
Not Autoclavable |
Removable lid | |
EVA | |
1 | |
4 L |