Learn More
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates is a reusable EVA sealing cap that fits onto the top of the plate to retain the sample in the well during transit or to stop the well and its contents from becoming contaminated.
Brand: Fisherbrand™
Code : Z4
Additional Details : Weight : 0.79560kg
Description
- For friction sealing of plates during transport or storage
- Cap mats are an economic alternative to thermal or adhesive seals
- Cap mats can be sterilized and reused
- Constructed of high quality ethylene-vinyl acetate (EVA)
- Rnase/Dnase free
Specifications
White | |
Non-sterile | |
50 | |
Square |
EVA | |
96 | |
96 Well Square Sealing Cap | |
96-well Square Deep Microplates (MPN 219009, 219030, 219008, 219006) |