missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Evans Blue
£25.90 - £78.10
Chemical Identifiers
CAS | 314-13-6 |
---|---|
Molecular Formula | C34H24N6Na4O14S4 |
Molecular Weight (g/mol) | 960.79 |
MDL Number | MFCD00004021 |
InChI Key | KBNIFDASRCWYGC-GXNXWABVSA-J |
Synonym | C.I. 23860, Direct Blue 53 |
PubChem CID | 24832074 |
IUPAC Name | tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate |
SMILES | [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206640
|
Thermo Scientific Chemicals
195550050 |
5 g | Glass bottle |
£25.90
5g |
In Stock
Log in to see stock. |
||||
10656402
|
Thermo Scientific Chemicals
195550250 |
25 g | Glass bottle |
£78.10
25g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
314-13-6 | |
960.79 | |
KBNIFDASRCWYGC-GXNXWABVSA-J | |
24832074 | |
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 |
C34H24N6Na4O14S4 | |
MFCD00004021 | |
C.I. 23860, Direct Blue 53 | |
tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate |
Specifications
314-13-6 | |
MFCD00004021 | |
Solubility in water: 280g/L (20°C). Other solubilities: soluble in alcohol, acids, alkalies, insoluble in ether | |
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 | |
960.79 | |
960.78 |
C34H24N6Na4O14S4 | |
C.I. 23860, Direct Blue 53 | |
KBNIFDASRCWYGC-GXNXWABVSA-J | |
tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate | |
24832074 | |
Evans Blue |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Evans Blue
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only