missing translation for 'onlineSavingsMsg'
Learn More

Evans Blue tetrasodium salt, Tocris Bioscience™

Product Code. 18035152
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dye for assessing cell viability and blood brain barrier permeability; also EAAT inhibitor and iGluR antagonist

Potent inhibitor of L-glutamate uptake into synaptic vesicles. Also inhibits AMPA and kainate receptor-mediated currents (IC50 values are 220 and 150 nM respectively). P2X-selective purinoceptor antagonist. Also a non-permeating dye, stains damaged plasma membrane and enters in the cytoplasm and nucleus, used for checking cell viability.

Specifications

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
MDL Number MFCD00004021
Quantity 1 g
Synonym evans blue, evansblue, azovan blue, modr evansova czech, blekit evansa polish, evans blue, sodium salt, modr prima 53 czech, ccris 167, c.i. direct blue 53, 1,3-naphthalenedisulfonic acid, 6,6'-3,3'-dimethyl 1,1'-biphenyl-4,4'-diyl bis azo bis 4-amino-5-hydroxy-, tetrasodium salt
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
IUPAC Name tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
Molecular Weight (g/mol) 960.79
PubChem CID 24832074
Formula Weight 960.82
Percent Purity >95%
Chemical Name or Material Evans Blue tetrasodium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.