Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£35.75 - £162.00
Product Code | Brand | Standard Taper Joint Side | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint Side | Price | Quantity & Availability | |||||
11953381
|
Lenz Laborglasintrumente
03041410 |
NS 14/23 |
£162.00
Pack of 10 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
11886104
|
Lenz Laborglasintrumente
03042949 |
NS 29/32 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Especially for rotary evaporators
- With center neck and conical joint
- Pear-shaped
- With inscription label
- Material: DURAN™
Specifications
Duran Glass |