Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£77.75 - £216.00
Specifications
Material Duran Glass
View More Specs
Products 5
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11903391
View Documents
Lenz Laborglasintrumente
03044037
100 mL
£82.80
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
15695585
View Documents
Lenz Laborglasintrumente
03044049
250 mL
£103.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
11943391
View Documents
Lenz Laborglasintrumente
03044073
2 L
£177.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
11953391
View Documents
Lenz Laborglasintrumente
03044076
3 L
£216.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
11993381
View Documents
Lenz Laborglasintrumente
03044028
50 mL
£77.75
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Description
Description
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifications
Specifications
Duran Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating